Biochanin A ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5,7-Dihydroxy-3-(4-methoxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-methoxyphenyl)chromen-4-one, 4'-Methoxy-5,7-dihydroxyisoflavone, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-4'-methoxyisoflavone

B4098-200MG B4098-1G B4098-5G
TCIAB4098-200MGEA 279 DKK
TCIAB4098-200MG TCIAB4098-1G TCIAB4098-5G
Biochanin A ≥98.0% (by HPLC, titration analysis)
Biochanin A
Formel: C₁₆H₁₂O₅
MDL Number: MFCD00006839
CAS-nummer: 491-80-5
EINECS: 207-744-7

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 215.0 to 219.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR