N-Benzyl-N,N'-dimethylethylenediamine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B3246-25G B3246-5G
TCIAB3246-25GEA 3430 DKK
TCIAB3246-25G TCIAB3246-5G
N-Benzyl-N,N'-dimethylethylenediamine ≥98.0% (by GC, titration analysis)
N-Benzyl-N,N'-dimethylethylenediamine
Formel: C₁₁H₁₈N₂
Kogepunkt: 74°C
MDL Number: MFCD00014856
CAS-nummer: 102-11-4
EINECS: 203-006-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=292b5317f8f47cbf8bbde165a4f9c14402f7a9a5&vid=292b5317f8f47cbf8bbde165a4f9c14402f7a9a5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR