X-Gal ≥98.0% (by HPLC, total nitrogen) for biochemical research

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

B3201-1G B3201-200MG
TCIAB3201-1GEA 1030 DKK
TCIAB3201-1G TCIAB3201-200MG
X-Gal ≥98.0% (by HPLC, total nitrogen) for biochemical research
X-Gal
Formel: C₁₄H₁₅BrClNO₆
Molekylvægt: 408.63 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00005666
CAS-nummer: 7240-90-6
EINECS: 230-640-8
Merck Index: 14,10074
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e1806f115e9c07e1525fd20382162b45676c18e&vid=451d1910e04b462eaea9f399e990b57fa3e679d8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -60.0 to -72.0 deg (C=1, DMF)
beta-Galactosidase detection test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR