ω-Brom-4-methylacetophenon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: α-Bromo-4-methylacetophenone, Bromomethyl p-methylphenyl ketone, 2-Bromo-4'-methylacetophenone, 2-Bromo-1-(p-tolyl)ethanone, 4-Methylphenacyl bromide

B2107-25G B2107-5G
TCIAB2107-25GEA 862 DKK
TCIAB2107-25G TCIAB2107-5G
ω-Brom-4-methylacetophenon ≥98.0% (by GC)
ω-Brom-4-methylacetophenon
Formel: C₉H₉BrO
Kogepunkt: 159°C
Smeltepunkt: 95…97 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00000203
CAS-nummer: 619-41-0
EINECS: 210-595-0
UN nummer: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=738c243b875b6e5f93cc7e3c4a7aef84c6a3788e&vid=738c243b875b6e5f93cc7e3c4a7aef84c6a3788e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 55.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR