trans-2-Hexenyl butyrate ≥93.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Butyric Acid trans-2-Hexenyl Ester

B1075-25ML
TCIAB1075-25MLEA 542 DKK
TCIAB1075-25ML
trans-2-Hexenyl butyrate ≥93.0% (by GC)
trans-2-Hexenyl butyrate
Formel: C₁₀H₁₈O₂
Molekylvægt: 170.25 g/mol
Flammepunkt: 86°C
MDL Number: MFCD00036546
CAS-nummer: 53398-83-7
EINECS: 258-515-3

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR