1,2-Epoxybutan ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,2-Butylene oxide, 1,2-Butylene Oxide, 1,2-Epoxybutane, Ethylethylene Oxide, Ethyloxirane

B0719-25ML B0719-500ML B0719-100ML
TCIAB0719-25MLEA 148 DKK
TCIAB0719-25ML TCIAB0719-500ML TCIAB0719-100ML
1,2-Epoxybutan ≥99.0% (by GC)
1,2-Epoxybutane
Formel: C₄H₈O
Kogepunkt: 63 °C (1013 hPa)
Smeltepunkt: –130 °C
Massefylde: 0,83 g/cm³ (20 °C)
Flammepunkt: -22°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005153
CAS-nummer: 106-88-7
EINECS: 203-438-2
UN nummer: 3022
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff51ece749bb292a8923432b669479843963af95&vid=ff51ece749bb292a8923432b669479843963af95&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR