2-Ethylhexyl butyrate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Butyric acid 2-ethylhexyl ester, Butyric acid octyl ester

B0130-25ML B0130-500ML
TCIAB0130-25MLEA 263 DKK
TCIAB0130-25ML TCIAB0130-500ML
2-Ethylhexyl butyrate ≥98.0% (by GC)
2-Ethylhexyl butyrate
Formel: C₁₂H₂₄O₂
Kogepunkt: 237°C
MDL Number: MFCD00059428
CAS-nummer: 25415-84-3
EINECS: 246-959-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR