Sodium benzohydroxamate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Benzohydroxamic acid sodium salt

B0112-25G
TCIAB0112-25GEA 3160 DKK
TCIAB0112-25G
Sodium benzohydroxamate ≥98.0% (by HPLC, titration analysis)
Sodium benzohydroxamate
Formel: C₇H₇NO₂
MDL Number: MFCD00058967
CAS-nummer: 22513-32-2

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR