3-Amino-2,2-dimethyl-1-propanol ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A0914-25G A0914-5G
TCIAA0914-25GEA 1460 DKK
TCIAA0914-25G TCIAA0914-5G
3-Amino-2,2-dimethyl-1-propanol ≥97.0% (by GC, titration analysis)
3-Amino-2,2-dimethyl-1-propanol
Formel: C₅H₁₃NO
MDL Number: MFCD00059847
CAS-nummer: 26734-09-8
EINECS: 247-948-3
UN nummer: 3263
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b629ad7b649116fe1a9ca0b1769faff6fb2021bf&vid=b629ad7b649116fe1a9ca0b1769faff6fb2021bf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR