(1-Pentyl)triphenylphosphoniumbromid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Pentyltriphenylphosphonium bromide

A0862-25G
TCIAA0862-25GEA 345 DKK
TCIAA0862-25G
(1-Pentyl)triphenylphosphoniumbromid ≥98.0% (by HPLC, titration analysis)
(1-Pentyl)triphenylphosphoniumbromid
Formel: H₃C(CH₂)₄P(C₆H₅)₃Br
Smeltepunkt: 168 °C
MDL Number: MFCD00031630
CAS-nummer: 21406-61-1
EINECS: 244-374-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dbd9dfedd1a753560066aa055d157a3d50553ef&vid=2dbd9dfedd1a753560066aa055d157a3d50553ef&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 164.0 to 168.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR