2-(2-Aminoethoxy)ethanol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A0301-25ML A0301-500ML
TCIAA0301-25MLEA 140 DKK
TCIAA0301-25ML TCIAA0301-500ML
2-(2-Aminoethoxy)ethanol ≥98.0% (by GC, titration analysis)
2-(2-Aminoethoxy)ethanol
Formel: C₄H₁₁NO₂
Kogepunkt: 223°C
Smeltepunkt: –11 °C
Massefylde: 1,0966 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008181
CAS-nummer: 929-06-6
EINECS: 213-195-4
UN nummer: 3055
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9323e5c81bef779021a841cb525d19e6b991c18&vid=c9323e5c81bef779021a841cb525d19e6b991c18&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR