Cerex® HPSAX 96 Well Plate RCH
Leverandør: Tecan
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
412-0401RCH
TCAT412-0401RCHEA 0 DKK
TCAT412-0401RCH
Cerex® HPSAX 96 Well Plate RCH
Mikroplader Solid Phase Ekstraktions plader
Polymeric strong anionic exchange chemistry.
  • Ion-exchange
  • Liquid handler compatible
20 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in fast and uniform sample prep. Ideal for acidic compounds.