Kieselsyre magnesiumsalt for chromatography

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Kieselsyre magnesiumsalt, aktiveret, Florisil, Magnesiumsilikat

20281
SUPL20281EA 1850 DKK
SUPL20281
Kieselsyre magnesiumsalt for chromatography
Kieselsyre magnesiumsalt
Formel: MgO₃Si
Molekylvægt: 100.38 g/mol
MDL Number: MFCD00078249
CAS-nummer: 1343-88-0
EINECS: 215-681-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54f0d27f7de4b6e6c91eea6b029d494a121d7989&vid=54f0d27f7de4b6e6c91eea6b029d494a121d7989&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR