Cole-Parmer® HP-200 Analogue Hotplate with PTFE Platform
Leverandør: Cole Parmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
04801-08
STUA04801-08EA 42270 DKK
STUA04801-08
Cole-Parmer® HP-200 Analogue Hotplate with PTFE Platform
Varmeapparater Varmeplader
Impervious to chemical attack, even concentrated acids.
  • Specifically designed for acid digestions and trace metal analysis
  • Separate temperature controller allows safe placement outside the fume hood
  • Large, square top plate accommodates one large vessel or several smaller ones
  • Plate is rated IP 43; controller is rated IP 30
With a glass ceramic heating plate mounted in a solid block of pure PTFE, this rugged hotplate withstands boiling acid solutions and aggressive cleaning agents. The separate analogue temperature controller is connected via a 2 mMore Product Information
Bestil nu
Specifikationer
Elektriske specifikationer 230 VAC, 50/60 Hz
Environmentally Preferable
Model HP-200-AR