3-Mercapto-1,2-propanediol ≥97%

Leverandør: Spectrum Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: alpha-Thioglycerol, α-Monothioglycerol, Monothioglycerol, (±)-3-Mercapto-1,2-dihydroxy propane, 1-Thioglycerol, (±)-α-Monothioglycerine, (±)-1-Thioglycerine, (±)-3-Mercapto-1,2-propanediol

M1177-25ML
SPCMM1177-25MLEA 2380 DKK
SPCMM1177-25ML
3-Mercapto-1,2-propanediol ≥97%
3-Mercapto-1,2-propandiol

Monothioglycerol, also known as 3-Mercaptopropane-1,2-diol and thioglycerol, is often used as a matrix in liquid secondary ion mass spectrometry and fast atom bombardment mass spectrometry.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₃H₈O₂S
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00004879
CAS-nummer: 96-27-5
UN nummer: 2810
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Assay (Titration By Iodine) 97% min.
Appearance Clear, Colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR