Choline chloride USP dietary supplement

Leverandør: Spectrum Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-hydroxy-N,N,N-trimethylethanaminium chloride

C2058-2.5KG C2058-500GM
SPCMC2058-2.5KGEA 6000 DKK
SPCMC2058-2.5KG SPCMC2058-500GM
Choline chloride USP dietary supplement
Choline chloride

Choline Chloride, USP Dietary Supplement.


All Spectrum Chemical USP Dietary Supplement grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Formel: HOH₂CCH₂N(CH₃)₃Cl
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011721
CAS-nummer: 67-48-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b015e76f45084bc46da3249578a85652cd6802&vid=85c8a0a1b152b861b3bc3ec8fe25a70509221d85&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR