Benzylbenzoat USP

Leverandør: Spectrum Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Benzoic acid benzyl ester

BE159-200LTBL BE159-20LT BE159-500ML
SPCMBE159-200LTBLEA 0 DKK
SPCMBE159-200LTBL SPCMBE159-20LT SPCMBE159-500ML
Benzylbenzoat USP
Benzylbenzoat

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formel: C₁₄H₁₂O₂
Kogepunkt: 323 °C (1013 hPa)
Smeltepunkt: 17…20 °C
Massefylde: 1,118 g/cm³ (20 °C)
Flammepunkt: 158 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003075
CAS-nummer: 120-51-4
EINECS: 204-402-9
UN nummer: 3082
ADR: 9,III
Merck Index: 13,01129

Bestil nu

Specifikationer Testresultater

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR