8-Anilino-1-naphthalenesulphonic acid ≥95%

Leverandør: Spectrum Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid

A1297-2.5KGBL A1297-5KGBL
SPCMA1297-2.5KGBLEA 53410 DKK
SPCMA1297-2.5KGBL SPCMA1297-5KGBL
8-Anilino-1-naphthalenesulphonic acid ≥95%
8-Anilino-1-naphthalensulfonsyre

8-Anilino-1-naphthalenesulfonic Acid is also known as ANS contains an amine group and a sulfonic acid. 8-Anilino-1-naphthalenesulfonic acid is a fluorescent dye with high affinity towards hydrophobic surfaces of proteins. It is useful for determining the affinity of hydrophobic ligands to their corresponding binding proteins and is used as a probe to monitor protein-folding.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₁₆H₁₃NO₃S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003998
CAS-nummer: 82-76-8

Bestil nu

Specifikationer Testresultater

Assay 95% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR