Concrete moisture analyser
Leverandør: Industrial Physics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LI9200
SIMELI9200EA 3810 DKK
SIMELI9200
Concrete moisture analyser
Analysatorer Fugtanalysatorer
This meter is a non-destructive moisture meter for concrete. By measuring the electrical impedance, the moisture content of concrete can be determined. Readings are taken by pressing the instrument against the surface.
  • Quick testing of large concrete floors or constructions
  • Automatic turn-off option
  • Four scales with normal and maximum 'HOLD' mode
The electrical impedance is measured by generating a low frequency electric field between the eight electrodes. Depending on the moisture content, the measurements are taken at a depth of several centimetres.