CRS ZPure™ O₂/H₂O Gas Filters
Leverandør: Trajan Scientific and Medical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
202295-B 202295D-B 202295D-SS 202295L-B 202295L-SS 202295-SS 202295XL-B 202295XL-SS 202296-B 202296D-B 202296D-SS 202296L-B 202296L-SS 202296-SS 202296XL-B 202296XL-SS 202295-B-QC 202295D-B-QC 202295D-SS-QC 202295L-B-QC 202295L-SS-QC 202295-SS-QC 202295XL-B-QC 202295XL-SS-QC 202296-B-QC 202296D-B-QC 202296D-SS-QC 202296L-B-QC 202296L-SS-QC 202296-SS-QC 202296XL-B-QC 202296XL-SS-QC 202RO2-B-QC 202RO2-SS-QC 202ROH-B-QC 202ROHD-B-QC 202ROHD-SS-QC 202ROHL-B-QC 202ROHL-SS-QC 202ROH-SS-QC 202ROHXL-B-QC 202ROHXL-SS-QC
SGEA202295-BEA 1900 DKK
SGEA202295-B SGEA202295D-B SGEA202295D-SS SGEA202295L-B SGEA202295L-SS SGEA202295-SS SGEA202295XL-B SGEA202295XL-SS SGEA202296-B SGEA202296D-B SGEA202296D-SS SGEA202296L-B SGEA202296L-SS SGEA202296-SS SGEA202296XL-B SGEA202296XL-SS SGEA202295-B-QC SGEA202295D-B-QC SGEA202295D-SS-QC SGEA202295L-B-QC SGEA202295L-SS-QC SGEA202295-SS-QC SGEA202295XL-B-QC SGEA202295XL-SS-QC SGEA202296-B-QC SGEA202296D-B-QC SGEA202296D-SS-QC SGEA202296L-B-QC SGEA202296L-SS-QC SGEA202296-SS-QC SGEA202296XL-B-QC SGEA202296XL-SS-QC SGEA202RO2-B-QC SGEA202RO2-SS-QC SGEA202ROH-B-QC SGEA202ROHD-B-QC SGEA202ROHD-SS-QC SGEA202ROHL-B-QC SGEA202ROHL-SS-QC SGEA202ROH-SS-QC SGEA202ROHXL-B-QC SGEA202ROHXL-SS-QC
CRS ZPure™ O₂/H₂O Gas Filters
Filtre Gasfiltre
ZPure™ O₂/H₂O gas filters removes oxygen and water from inert gases, He, Ar, N₂ and H₂, making it ideal for use with GC and GC/MS carrier gas lines. It differs from the Zpure dual filter in that it is completely non-hazardous and is recommended for any application requiring ultra-pure gas, free from oxygen and water.
  • High efficiency in-line traps with outstanding capacity
  • High quality activated adsorbents for long purifier life and efficient contaminant removal
  • Various size and fitting configurations to fit existing installations
  • Filter lifetime is