Urea

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Urinstof

39305.01
SERA39305.01EA 238.85 DKK
SERA39305.01
Urea
Urea
Formel: NH₂CONH₂
Kogepunkt: 197 °C (1013 hPa)
Massefylde: 1,34 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008022
CAS-nummer: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR