Precast horizontal gel kits, SERVALYT™ PRECOTES™ IEF Starter Kit
Leverandør: Serva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
39060.01
SERA39060.01EA 1821.25 DKK
SERA39060.01
Precast horizontal gel kits, SERVALYT™ PRECOTES™ IEF Starter Kit
Elektroforese geler Forstøbte Geler
SERVALYT™ PRECOTES™ are easy to start, ready to use kits for special applications.
SERVALYT™ PRECOTES™ IEF Starter Kit contains:
3× SERVALYT™ PRECOTES™ 125×125 mm with PAG layer 300 µm
20× electrode wicks
1× applicator strips
2×10 ml electrode buffer solutions
10 ml heat exchange liquid
100 mg SERVA Blue W
100 mg SERVA Violet 17
60 µl IEF marker 3-10 SERVA liquid mix