4,4'-Bi-o-toluidin stabilised analytical reagent

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4,4'-Bi-o-toluidine

Stabilization: Moistened with water

36689.01 36689.02
SERA36689.01EA 0 DKK
SERA36689.01 SERA36689.02
4,4'-Bi-o-toluidin stabilised analytical reagent
4,4'-Bi-o-toluidin
Formel: [-C₆H₃(CH₃)-₄-NH₂]₂
Molekylvægt: 212.29 g/mol
Kogepunkt: ∼361 °C (1013 hPa)
Smeltepunkt: 129…131 °C
Massefylde: ∼1.234 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00014773
CAS-nummer: 119-93-7
EINECS: 204-358-0
UN nummer: 3077
ADR: 9 III
Merck Index: 12,09651

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR