EDTA

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II

11278.01 11278.02
SERA11278.01EA 156.75 DKK
SERA11278.01 SERA11278.02
EDTA
EDTA
Formel: C₁₀H₁₆N₂O₈
Kogepunkt: 614 °C (1013 hPa)
Smeltepunkt: 220 °C
Massefylde: 0,86 g/cm³ (20 °C)
Flammepunkt: >100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003541
CAS-nummer: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR