Acrylamid-Bisacrylamid (37.5:1) 40% in aqueous solution

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Acrylamid-Bis

10681.02 10681.03
SERA10681.02EA 1022.58 DKK
SERA10681.02 SERA10681.03
Acrylamid-Bisacrylamid (37.5:1) 40% in aqueous solution
Acrylamid-Bis (32:1)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00080848
UN nummer: 3426
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR