Tilbehør til SCAT SafetyCaps
Leverandør: S.C.A.T.
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
160506 107044 107045 107620 107802 107803 107804 107806 107807 107808 107810 107814 107816 107817 107820 107823 107824 117816 107046 107047 160501 160502 106490 107680 160523 350110 450110
590-0826EA 173 DKK
590-0826 590-0528 590-0529 590-0555 590-0580 590-0581 590-0582 590-0584 590-0585 590-0586 590-0587 590-0591 590-0593 590-0594 590-0597 590-0600 590-0601 554-4730 SCTV107046 SCTV107047 554-2211 SCTV160502 SCTV106490 SCTV107680 SCTV160523 SCTV350110 SCTV450110
Tilbehør til SCAT SafetyCaps
Låg Sikkerhedslåg og sikkerhedsaffaldslåg
Helpful supplies for the safe use of SCAT and SymLine products Safety Caps and Safety Waste Caps.
  • SCAT connectors are available in the following versions:
  • Conical connectors
  • Y-connectors
  • Tube connectors
  • Blind plugs
  • Capillary connector
SCAT thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters, or other disposal and supply units.