Stopcock, three-way, T-bore, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
286416405 286419805 286417804 286418406
SCOT286416405EA 1620 DKK
SCOT286416405 SCOT286419805 SCOT286417804 SCOT286418406
Stopcock, three-way, T-bore, DURAN®
Stophaner
DURAN®, borosilicate glass 3.3, clear, with solid SBW glass or PTFE key.
  • Replaceable NS keys with T-bore
  • Screw thread retaining device