Stopcock keys for burettes, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
286035504
SCOT286035504EA 145 DKK
SCOT286035504
Stopcock keys for burettes, DURAN®
Stophane nøgler
DURAN®, borosilicate glass 3.3, clear.
  • Side entry
  • Drilled and grounded
  • With three part retaining device