BioDrop Duo+ Microvolume Spectrophotometer
Leverandør: BIOCHROM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
80-3006-68
SCLI80-3006-68EA 0 DKK
SCLI80-3006-68
BioDrop Duo+ Microvolume Spectrophotometer
Spektrofotometer og fotometer Biofotometre
BioDrop's unique sample port is dedicated to microvolume measurement. The port is easy to use: simply pipette as little as 0,5 μl and measure. Cleaning the port is easy too, just wipe with lint-free tissue to reduce sample carryover to undetectable amounts. BioDropDuo+ also provides a standard cuvette port.
  • The in-built sample port uses no moving parts
  • Instrument provides excellent reproducibility without the need to recondition or calibrate
  • Highly accurate with the pathlength of the port specified to ±5 μm
  • Export data via USB or link to PC