NMR tubes, polyethylene pressure caps
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SP897095-0001 SP897095-0021 SP897095-0061 SP897095-0071 SP897095-0081 SP897095-1001
SCERSP897095-0001EA 231 DKK
SCERSP897095-0001 SCERSP897095-0021 SCERSP897095-0061 SCERSP897095-0071 SCERSP897095-0081 SCERSP897095-1001
NMR tubes, polyethylene pressure caps
Låg Reagensglaslåg
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporisation pressures.