Chromatography columns, Flex-Column®, KIMBLE®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SP420400-0710 SP420400-2510 SP420401-1020 SP420401-1030 SP420401-2510 SP420400-1520 SP420400-2520 SP420400-1530 SP420401-0715 SP420400-2575 SP420401-1005 SP420401-1011 SP420400-2530 SP420400-0720 SP420401-1530 SP420400-1020 SP420400-1510 SP420400-1030 SP420400-1015 SP420401-0720 SP420401-0730 SP420401-2520 SP420401-2530 SP420401-2515
SCERSP420400-0710EA 0 DKK
SCERSP420400-0710 SCERSP420400-2510 SCERSP420401-1020 SCERSP420401-1030 SCERSP420401-2510 SCERSP420400-1520 SCERSP420400-2520 SCERSP420400-1530 SCERSP420401-0715 SCERSP420400-2575 SCERSP420401-1005 SCERSP420401-1011 SCERSP420400-2530 SCERSP420400-0720 SCERSP420401-1530 SCERSP420400-1020 SCERSP420400-1510 SCERSP420400-1030 SCERSP420400-1015 SCERSP420401-0720 SCERSP420401-0730 SCERSP420401-2520 SCERSP420401-2530 SCERSP420401-2515
Chromatography columns, Flex-Column®, KIMBLE®
Kromatografikolonner
These economical columns are constructed of polypropylene reservoirs and column outlets permanently connected to a borosilicate glass barrel, molded PP end caps, and HDPE bed supports. They also feature Luer lock inlet and outlet fittings for better connections.
  • Easy connection to any low pressure chromatography system
  • Ideal for use with gel filtration, ion exchange, affinity and adsorption media
PP packing reservoir funnels are for use in initial column packing or as small buffer reservoirs. Glass packing reservoirs are ideal for packing gel slurriesMore Product Information