Containers, disposable, with screw cap
Leverandør: SARSTEDT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
75.563 75.562.105 76.564 75.562.005 75.562.300
SARS75.563EA 417.92 DKK
SARS75.563 SARS75.562.105 SARS76.564 SARS75.562.005 SARS75.562.300
Containers, disposable, with screw cap
Prøvecontainere Allround kasser
PP, translucent, with neutral HDPE screw cap
  • Sterile or non-sterile
  • Without lid or with attached or enclosed lid available
  • Safety label ensures sterility prior to use and provides a convenient writing space
Ideal as general specimen containers.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904