Microtubes with thread in special sterile packaging
Leverandør: SARSTEDT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
72.607.772 72.609.711 72.664.711 72.687.772 72.730.711
SARS72.607.772EA 0 DKK
SARS72.607.772 SARS72.609.711 SARS72.664.711 SARS72.687.772 SARS72.730.711
Microtubes with thread in special sterile packaging
Rør Mikrorør
PP, translucent
  • With or without skirted base
Each bag is sub-packaged into a second additional bag. Removing the outer bag leaves a complete and sterile bag packaging for convenient processing in clean rooms or sterile environments. This packaging option is ideal for users working under clean room conditions to fill precious reagents into tubes.