Capillary tubes
Leverandør: SARSTEDT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
19.447.001
SARS19.447.001EA 0 DKK
SARS19.447.001
Capillary tubes
Kliniske diagnose systemer Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Glass, transparent.
  • End-to-end capillaries
  • Prepared with potassium EDTA