Antibacterial Foam Soaps
Leverandør: Rubbermaid Commercial Products
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2167858 2167901 2167860 2167904
RUBB2167858EA 293 DKK
RUBB2167858 RUBB2167901 RUBB2167860 RUBB2167904
Antibacterial Foam Soaps
Hudpleje Sæber og huden rensemidler
Antibacterial soap kills germs, which is ideal for facilities looking for added peace of mind.
  • Kills 99,99% of common germs; fully tested and proven to be bactericidal, virucidal, and yeasticidal
  • Broad spectrum activity disinfects hands in 30 seconds
  • Fragrance-free; non-tainting for food environments
  • Pleasant to use, rich foam formula is ideal for any location
This fast-acting, fragrance-free solution contains moisturising ingredients to help prevent skin dryness. The biodegradable formula supports environmental wellbeing. Controlled dispensingMore Product Information