Natriumazid 0,05% water, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Sodium triazide

RTC000068-1L
RTC000068-1LEA 612 DKK
RTC000068-1L
Natriumazid 0,05% water, Supelco®
Sodium azide
Formel: NaN₃
Molekylvægt: 65.01 g/mol
Smeltepunkt: 275
MDL Number: MFCD00003536
CAS-nummer: 26628-22-8
Merck Index: 13,08653

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR