Trådkurve, Remanit® 4301
Leverandør: RSG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
120.010 120.012 120.014 120.016 120.018 120.020 120.030 120.040
RSGA120.010EA 468 DKK
RSGA120.010 RSGA120.012 RSGA120.014 RSGA120.016 RSGA120.018 RSGA120.020 RSGA120.030 RSGA120.040
Trådkurve, Remanit® 4301
Kurve Kurve, tråd
These electropolished wire baskets are made of stainless steel ¹⁸/₁₀.
  • Very good temperature resistance
  • Good chemical resistance
  • Reinforced edges