Mangan(II)chlorid tetrahydrat

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate

T881.1 T881.2
ROTHT881.1EA 384.4 DKK
ROTHT881.1 ROTHT881.2
Mangan(II)chlorid tetrahydrat
Mangan(II)chlorid tetrahydrat
Formel: MnCl₂·4H₂O
Molekylvægt: 197.9 g/mol
Kogepunkt: 1190
Smeltepunkt: 58…59 °C
Massefylde: 2.01 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149792
CAS-nummer: 13446-34-9
EINECS: 231-869-6
UN nummer: 3077
ADR: 9 III
Merck Index: 13,05751

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR