Metylorange 0,1% aqueous solution

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

T119.1
ROTHT119.1EA 185.72 DKK
ROTHT119.1
Metylorange 0,1% aqueous solution
Metylorange
Formel: C₁₄H₁₄N₃NaO₃S
Molekylvægt: 327.34 g/mol
Kogepunkt: 90
Flammepunkt: 38
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007502
CAS-nummer: 547-58-0
EINECS: 208-925-3
Merck Index: 13,06127

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR