ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: NADPH tetrasodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1(4H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)oxidophosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl phosphate, Nicotinamide adenine dinucleotide phosphate tetrasodium salt, (reduced form)

AE14.1 AE14.2
ROTHAE14.1EA 745.67 DKK
ROTHAE14.1 ROTHAE14.2
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
Formel: C₂₁H₂₆N₇Na₄O₁₇P₃
MDL Number: MFCD00036263
CAS-nummer: 2646-71-1
EINECS: 220-163-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR