Diethylether

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ether

AE04.1
ROTHAE04.1EA 447.1 DKK
ROTHAE04.1
Diethylether
Diethylether
Formel: (CH₃CH₂)₂O
Kogepunkt: 35 °C (1013 hPa)
Smeltepunkt: –123 °C
Massefylde: 0,7135 g/cm³ (20 °C)
Flammepunkt: –40 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011646
CAS-nummer: 60-29-7
EINECS: 200-467-2
UN nummer: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR