(±)-2-Hexanol

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane

9463.1
ROTH9463.1EA 0 DKK
ROTH9463.1
(±)-2-Hexanol
(±)-2-Hexanol
Formel: C₆H₁₄O
Molekylvægt: 102.18 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004585
CAS-nummer: 626-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9db7ec887376d519b077785cdead15c1aed20d58&vid=5c0c76fb523ec80ddb5134157d5d19ce7a63dcc3&prid=web&clid=SAPDAM

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR