L-Threonin

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid

7534.1
ROTH7534.1EA 0 DKK
ROTH7534.1
L-Threonin
L-Threonin
Formel: C₄H₉NO₃
Molekylvægt: 119.12 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064270
CAS-nummer: 72-19-5
EINECS: 200-774-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR