Kulstof aktiveret

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Carbon activated, Activated carbon, Activated charcoal

5966.1 5966.2 5966.4
ROTH5966.1EA 0 DKK
ROTH5966.1 ROTH5966.2 ROTH5966.4
Kulstof aktiveret
Kulstof aktiveret
Formel: C
Smeltepunkt: 3550 °C
Massefylde: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS-nummer: 7440-44-0
EINECS: 231-153-3
UN nummer: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR