Methyl 4-hydroxybenzoat

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

4748.1
ROTH4748.1EA 0 DKK
ROTH4748.1
Methyl 4-hydroxybenzoat
Methyl 4-hydroxybenzoat
Formel: HOC₆H₄CO₂CH₃
Molekylvægt: 152.15 g/mol
Kogepunkt: 270…280 °C (1013 hPa)
Smeltepunkt: 125…128 °C
Massefylde: 1.1208 g/cm³ (137 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002352
CAS-nummer: 99-76-3
EINECS: 202-785-7
Merck Index: 13,06129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR