Dimethylphthalat

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Phthalic acid dimethyl ester

4576.1
ROTH4576.1EA 0 DKK
ROTH4576.1
Dimethylphthalat
Dimethylphthalat
Formel: C₁₀H₁₀O₄
Molekylvægt: 194.19 g/mol
Kogepunkt: 283.7
Smeltepunkt: 6
Massefylde: 1.192 g/cm³ (20 °C)
Flammepunkt: 146
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008425
CAS-nummer: 131-11-3
EINECS: 205-011-6
Merck Index: 13,03281

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR