Ammoniumcerium (IV)sulfat dihydrat

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Tetraammonium cerium tetrakis(sulphate) dihydrate, Cerium (IV) ammonium sulphate dihydrate, Tetraammonium cerium(IV) sulphate dihydrate, Ammonium cerium(IV) sulphate hydrate, Ammonium ceric sulphate dihydrate

3338.1
ROTH3338.1EA 0 DKK
ROTH3338.1
Ammoniumcerium (IV)sulfat dihydrat
Ammoniumcerium (IV)sulfat dihydrat
Formel: (NH₄)₂Ce(SO₄)₄·2H₂O
Smeltepunkt: 130 °C
MDL Number: MFCD00149417
CAS-nummer: 10378-47-9
EINECS: 231-567-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR