Safety transport containers
Leverandør: ROETZMEIER SICHERH BEHAELTER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
01T 02T 05T 05KT 10KT 20KTL 10TZ 25TZ 05KTV 10KTV
119-2350EA 1340 DKK
119-2350 119-2351 119-2352 119-2370 119-2371 119-0122 119-0157 119-0156 ROET05KTV 119-0209
Safety transport containers
Sikkerhedsdunke
Safety transport containers for UN approved transport of hazardous goods on public routes.
  • High quality, and chemically resistant stainless steel with PTFE gaskets
  • All types of hazardous goods (UN X approval)
  • Help to avoid non returnable one-way transport packaging
  • Screw cap without pressure control valve - also available with pressure control valve as well
  • Removable flame trap
Design approved according to ADR, RID, IMDG Code, ICAO-TI and GGVSEB.
Certificeringer: Design approved according to ADR, RID, IMDG Code, ICAO-TI and GGVSEB.