Buffers, TRIS HCl solutions, pH range 7,2 to 8,5 (1 mol/L)
Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MB-001 MB-002 MB-003 MB-004 MB-005 MB-027-1000
ROCKMB-001EA 1040 DKK
ROCKMB-001 ROCKMB-002 ROCKMB-003 ROCKMB-004 ROCKMB-005 ROCKMB-027-1000
Buffers, TRIS HCl solutions, pH range 7,2 to 8,5 (1 mol/L)
Buffere Biologiske buffere
Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.
These solutions were aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilized containers. The products were tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Advarsel: These products ares for research use only and are not intended for therapeutic or diagnostic applications.