Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

606-103-129
ROCK606-103-129EA 3700 DKK
ROCK606-103-129
Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)
Antistoffer
Anti-Guinea Pig IgG HRP is suitable for immunoblotting, western blot, dot blot, ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:100000 - 1:200,000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Guinea Pig
Bestil nu

Specifikationer

Antigen symbol IgG (H+L)
Environmentally Preferable
Antistof type Secondary
Clonality Polyclonal
Konjugation Peroxidase
Reactivity Guinea pig
Vært Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Gel skift Yes
Flowcytometri Yes
ChIP Yes
Neutralisation Yes
Immunogen Anti-Guinea Pig IgG (H&L) was produced by repeated immunization with Guinea Pig IgG whole molecule in goat.
Udformning Antibody fraction
Form Lyophilized
Purification purified by immunoaffinity chromatography
Koncentration 1,0 mg/ml by UV absorbance at 280 nm
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Opbevaringstemperatur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR